2-Brom-4-fluor-1-(methylsulfonyl)benzol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-4-fluorophenyl methyl sulphone
OT-1002-1G
OT-1002-5G
OT-1002-250M
COBBOT-1002-1GEA
423
EUR
COBBOT-1002-1G
COBBOT-1002-5G
COBBOT-1002-250M
2-Brom-4-fluor-1-(methylsulfonyl)benzol
2-Brom-4-fluor-1-(methylsulfonyl)benzol
Formel:
C₇H₆BrFO₂S Molecular Weight: 253.0887 g/mol |
MDL:
MFCD13195736 CAS-Nummer: 1039744-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...