2-Brom-N,N-diethyl-4-nitroanilin

Lieferant: COMBI-BLOCKS

HC-2682-5G HC-2682-1G
COBBHC-2682-5GEA 451 EUR
COBBHC-2682-5G COBBHC-2682-1G
2-Brom-N,N-diethyl-4-nitroanilin
2-Brom-N,N-diethyl-4-nitroanilin
Formel: C₁₀H₁₃BrN₂O₂
MDL: MFCD12026041
CAS-Nummer: 1150271-18-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR