2-Brom-N,N-dimethyl-4-nitroanilin

Lieferant: COMBI-BLOCKS

HC-2680-1G HC-2680-5G
COBBHC-2680-1GEA 102 EUR
COBBHC-2680-1G COBBHC-2680-5G
2-Brom-N,N-dimethyl-4-nitroanilin
2-Brom-N,N-dimethyl-4-nitroanilin
Formel: C₈H₉BrN₂O₂
Molecular Weight: 245.733 g/mol
MDL: MFCD00179577
CAS-Nummer: 64230-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR