5-Amino-2-bromo-4-nitrophenol
Lieferant: COMBI-BLOCKS
AN-1539-5G
AN-1539-1G
COBBAN-1539-5GEA
0
EUR
COBBAN-1539-5G
COBBAN-1539-1G
5-Amino-2-bromo-4-nitrophenol
5-Amino-2-bromo-4-nitrophenol
Formel:
C₆H₅BrN₂O₃ |
MDL:
MFCD16036100 CAS-Nummer: 855836-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...