2-Chlor-5-cyanphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8243-1G BB-8243-5G
COBBBB-8243-1GEA 147 EUR
COBBBB-8243-1G COBBBB-8243-5G
2-Chlor-5-cyanphenylboronsäure
2-Chlor-5-cyanphenylboronsäure
Formel: C₇H₅BClNO₂
Molecular Weight: 181,3841 g/mol
MDL: MFCD06656271
CAS-Nummer: 936249-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR