2-Chlor-5-cyanphenylboronsäure

Lieferant: FLUOROCHEM

016220-1G 016220-5G
FLUO016220-1GEA 149 EUR
FLUO016220-1G FLUO016220-5G
2-Chlor-5-cyanphenylboronsäure
2-Chlor-5-cyanphenylboronsäure
Formel: C₇H₅BClNO₂
Molecular Weight: 181.3841 g/mol
MDL: MFCD06656271
CAS-Nummer: 936249-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR