2-Acetamido-5-chlorphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-5084-1G
BB-5084-250M
COBBBB-5084-1GEA
0
EUR
COBBBB-5084-1G
COBBBB-5084-250M
2-Acetamido-5-chlorphenylboronsäure
2-Acetamido-5-chlorphenylboronsäure
Formel:
C₈H₉BClNO₃ |
MDL:
MFCD11504839 CAS-Nummer: 1072945-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...