5-Chlorsalicylamid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Chloro-2-hydroxybenzamide

L17897.14 L17897.06
L17897.14EA 70 EUR
L17897.14 L17897.06
5-Chlorsalicylamid ≥97%
5-Chlorsalicylamid

Purity: 97% CAS No.: 7120-43-6 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 223-226° Beilstein Reference: 743348

Formel: C₇H₆ClNO₂
Molecular Weight: 171.58 g/mol
Schmelzpunkt: 223…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007980
CAS-Nummer: 7120-43-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR