3-Chlor-2-fluor-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

HC-2565-1G HC-2565-5G
COBBHC-2565-1GEA 98.3 EUR
COBBHC-2565-1G COBBHC-2565-5G
3-Chlor-2-fluor-4-thiocyanatoanilin
3-Chlor-2-fluor-4-thiocyanatoanilin
Formel: C₇H₄ClFN₂S
MDL: MFCD12026028
CAS-Nummer: 329926-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR