1-Boc-4-(tosyloxy)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: 1-Boc-4-(Tosyloxy)piperidine, 1-(tert-Butoxycarbonyl)-4-(p-toluenesulphonyloxy)piperidine, 1-Boc-4-tosyloxypiperidine, 4-Tosyloxypiperidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)-4-piperidyl p-Toluenesulphonate, tert-Butyl 4-(p-Toluenesulphonyloxy)piperidine-1-carboxylate, tert-Butyl 4-Tosyloxypiperidine-1-carboxylate, 1-Boc-4-piperidyl p-Toluenesulphonate, 4-(p-Toluenesulphonyloxy)piperidine-1-carboxylic acid tert-Butyl ester

AM-1737-5G AM-1737-1G AM-1737-25G
COBBAM-1737-5GEA 76.7 EUR
COBBAM-1737-5G COBBAM-1737-1G COBBAM-1737-25G
1-Boc-4-(tosyloxy)piperidin
1-Boc-4-(tosyloxy)piperidin
Formel: C₁₇H₂₅NO₅S
Molecular Weight: 355.4500 g/mol
MDL: MFCD06796535
CAS-Nummer: 118811-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR