4-Methoxy-3-nitrobenzamid
Lieferant: Thermo Scientific
SPB07250.1
SPB07250.10
MAYBSPB07250.1EA
0
EUR
MAYBSPB07250.1
MAYBSPB07250.10
4-Methoxy-3-nitrobenzamid
4-Methoxy-3-nitrobenzamid
Formel:
C₈H₈N₂O₃ Molecular Weight: 180,17 g/mol |
MDL:
MFCD00017136 CAS-Nummer: 10397-58-7 EINECS: 233-859-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...