2-(4-Aminophenoxy)benzoesäure
Lieferant: BIONET RESEARCH
7Y-0701.1
7Y-0701.10
7Y-0701.5
7Y-0701.500
BIOT7Y-0701.1EA
0
EUR
BIOT7Y-0701.1
BIOT7Y-0701.10
BIOT7Y-0701.5
BIOT7Y-0701.500
2-(4-Aminophenoxy)benzoesäure
2-(4-Aminophenoxy)benzoic acid
Formel:
C₁₃H₁₁NO₃ |
MDL:
MFCD06659583 CAS-Nummer: 67724-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...