2-Methoxy-5-sulfamoylbenzoesäure ≥97%

Lieferant: Thermo Scientific
Achtung

B23698.09 B23698.18
B23698.09EA 21 EUR
B23698.09 B23698.18
2-Methoxy-5-sulfamoylbenzoesäure ≥97%
2-Methoxy-5-sulfamoylbenzoesäure

Purity: 97% CAS No.: 22117-85-7 Molecular Formula: C8H9NO5S Formula Weight: 232.23 MDL No.: MFCD00129997

Formel: C₈H₉NO₅S
Molecular Weight: 232,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00129997
CAS-Nummer: 22117-85-7
EINECS: 244-789-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR