3-(Chlorsulfonyl)benzoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

B22987.06 B22987.14
B22987.06EA 38.8 EUR
B22987.06 B22987.14
3-(Chlorsulfonyl)benzoesäure ≥97%
3-(Chlorsulfonyl)benzoesäure

Purity: 97% CAS No.: 4025-64-3 Molecular Formula: C7H5ClO4S Formula Weight: 220.63 Melting Point: 133-138° Beilstein Reference: 2645465 UN No.: 3261 MDL No.: MFCD00024877

Formel: C₇H₅ClO₄S
Molecular Weight: 220.63 g/mol
Schmelzpunkt: 133…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024877
CAS-Nummer: 4025-64-3
EINECS: 223-692-8
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR