3-(Chlorsulfonyl)benzoesäure

Lieferant: Thermo Scientific

MAYBBTB08648.10EA 0 EUR
MAYBBTB08648.10 MAYBBTB08648.50 MAYBBTB08648.1
3-(Chlorsulfonyl)benzoesäure
3-(Chlorsulfonyl)benzoesäure
Formel: C₇H₅ClO₄S
Molecular Weight: 220,63 g/mol
MDL: MFCD00024877
CAS-Nummer: 4025-64-3
EINECS: 223-692-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR