3-(Chlorsulfonyl)benzoesäure, Sigma-Aldrich®

Lieferant: Merck

441759-25G 441759-5G
SIAL441759-25GEA 112 EUR
SIAL441759-25G SIAL441759-5G
3-(Chlorsulfonyl)benzoesäure, Sigma-Aldrich®
3-(Chlorsulfonyl)benzoesäure
Formel: C₇H₅ClO₄S
Molecular Weight: 220.63 g/mol
MDL: MFCD00024877
CAS-Nummer: 4025-64-3
EINECS: 223-692-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR