4-(Chlorsulfonyl)benzoesäure 96%

Lieferant: Thermo Fisher Scientific
Gefahr

A17337.03 A17337.06 A17337.14
A17337.03EA 23.8 EUR
A17337.03 A17337.06 A17337.14
4-(Chlorsulfonyl)benzoesäure 96%
4-(Chlorsulfonyl)benzoesäure

Purity: 96% CAS No.: 10130-89-9 Molecular Formula: C7H5ClO4S Formula Weight: 220.63 Odour: Acrid Melting Point: 228-232° UN No.: 3261 MDL No.: MFCD00007448

Formel: C₇H₅ClO₄S
Molecular Weight: 220.63 g/mol
Schmelzpunkt: 228…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007448
CAS-Nummer: 10130-89-9
EINECS: 233-367-2
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR