2-Sulfobenzoesäure Monoammoniumsalz

Lieferant: FLUOROCHEM

Synonyme: 2-sulphobenzoic acid ammonium salt

001259-250G 001259-50G
FLUO001259-250GEA 0 EUR
FLUO001259-250G FLUO001259-50G
2-Sulfobenzoesäure Monoammoniumsalz
2-Sulfobenzoesäure Monoammoniumsalz
Formel: C₇H₉NO₅S
MDL: MFCD00040590
CAS-Nummer: 6939-89-5
EINECS: 230-077-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR