2,6-Difluor-(4-methoxycarbonyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 3,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
PN-5841-1G
PN-5841-250M
COBBPN-5841-1GEA
862
EUR
COBBPN-5841-1G
COBBPN-5841-250M
2,6-Difluor-(4-methoxycarbonyl)phenylboronsäurepinakolester
2,6-Difluor-(4-methoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₄H₁₇BF₂O₄ Molecular Weight: 298.09 g/mol |
MDL:
MFCD12546491 CAS-Nummer: 1218791-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...