2,3-Difluor-(5-methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3,4-difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

PN-5682-1G PN-5682-250M
COBBPN-5682-1GEA 1490 EUR
COBBPN-5682-1G COBBPN-5682-250M
2,3-Difluor-(5-methoxycarbonyl)phenylboronsäurepinakolester
2,3-Difluor-(5-methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₇BF₂O₄
Molecular Weight: 298.09 g/mol
MDL: MFCD12026086
CAS-Nummer: 1218791-19-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR