3-(tert-Butyldimethylsilyloxy)-5-(trifluormethoxy)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-5387-1G
PN-5387-250M
COBBPN-5387-1GEA
0
EUR
COBBPN-5387-1G
COBBPN-5387-250M
3-(tert-Butyldimethylsilyloxy)-5-(trifluormethoxy)phenylboronsäurepinakolester
3-(tert-Butyldimethylsilyloxy)-5-(trifluormethoxy)phenylboronsäurepinakolester
Formel:
C₁₉H₃₀BF₃O₄Si |
MDL:
MFCD12026067 CAS-Nummer: 1150271-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...