4-Fluor-3-(tetrahydropyran-2-yloxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Fluoro-3-(tetrahydro-2H-pyran-2-yloxy)phenylboronic acid
BB-5999-5G
BB-5999-1G
COBBBB-5999-5GEA
1490
EUR
COBBBB-5999-5G
COBBBB-5999-1G
4-Fluor-3-(tetrahydropyran-2-yloxy)phenylboronsäure
4-Fluor-3-(tetrahydropyran-2-yloxy)phenylboronsäure
Formel:
C₁₁H₁₄BFO₄ |
MDL:
MFCD13195650 CAS-Nummer: 1217501-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...