3-(tert-Butyldimethylsilyloxy)-2,4,6-trifluorphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-5289-5G
BB-5289-1G
COBBBB-5289-5GEA
0
EUR
COBBBB-5289-5G
COBBBB-5289-1G
3-(tert-Butyldimethylsilyloxy)-2,4,6-trifluorphenylboronsäure
3-(tert-Butyldimethylsilyloxy)-2,4,6-trifluorphenylboronsäure
Formel:
C₁₂H₁₈BF₃O₃Si |
MDL:
MFCD11053859 CAS-Nummer: 1072946-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...