5-(tert-Butyldimethylsilyloxy)-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-((tert-Butyldimethylsilyl)oxy)-2-fluorophenyl)boronic acid, 5-(t-Butyldimethylsilyloxy)-2-fluorophenylboronic acid

BB-5633-5G BB-5633-1G
COBBBB-5633-5GEA 1490 EUR
COBBBB-5633-5G COBBBB-5633-1G
5-(tert-Butyldimethylsilyloxy)-2-fluorphenylboronsäure
5-(tert-Butyldimethylsilyloxy)-2-fluorphenylboronsäure
Formel: C₁₂H₂₀BFO₃Si
Molecular Weight: 270.1800 g/mol
MDL: MFCD11855846
CAS-Nummer: 1150114-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR