N-Boc-2-fluoranilin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (2-fluorophenyl)carbamate

AN-1389-5G AN-1389-25G AN-1389-1G
COBBAN-1389-5GEA 0 EUR
COBBAN-1389-5G COBBAN-1389-25G COBBAN-1389-1G
N-Boc-2-fluoranilin
N-Boc-2-fluoranilin
Formel: C₁₁H₁₄FNO₂
MDL: MFCD04112559
CAS-Nummer: 98968-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR