N-Boc-4-fluoranilin
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl (4-fluorophenyl)carbamate, 4-Fluoroaniline, N-BOC protected
AN-2561-25G
AN-2561-5G
COBBAN-2561-25GEA
0
EUR
COBBAN-2561-25G
COBBAN-2561-5G
N-Boc-4-fluoranilin
N-Boc-4-fluoranilin
Formel:
C₁₁H₁₄FNO₂ |
MDL:
MFCD07127707 CAS-Nummer: 60144-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...