N,N-Dimethyl-2,6-difluoranilin

Lieferant: Thermo Scientific

SEW01802.250 SEW01802.1
MAYBSEW01802.250EA 0 EUR
MAYBSEW01802.250 MAYBSEW01802.1
N,N-Dimethyl-2,6-difluoranilin
N,N-Dimethyl-2,6-difluoranilin
Formel: C₈H₉F₂N
MDL: MFCD00832734
CAS-Nummer: 55847-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR