3,4-Difluor-2-methoxyanilinsulfat

Lieferant: COMBI-BLOCKS

AN-1364-5G AN-1364-1G
COBBAN-1364-5GEA 451 EUR
COBBAN-1364-5G COBBAN-1364-1G
3,4-Difluor-2-methoxyanilinsulfat
3,4-Difluor-2-methoxyanilinsulfat
Formel: C₇H₉F₂NO₅S
MDL: MFCD11053865
CAS-Nummer: 1072945-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR