2,5-Difluor-4-thiocyanatoanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1408-5G AN-1408-1G
COBBAN-1408-5GEA 303 EUR
COBBAN-1408-5G COBBAN-1408-1G
2,5-Difluor-4-thiocyanatoanilin Hydrochlorid
2,5-Difluor-4-thiocyanatoanilin Hydrochlorid
Formel: C₇H₅ClF₂N₂S
MDL: MFCD11855821
CAS-Nummer: 1150114-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR