4-Nitrophenethylamin Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
2-(4-Nitrophenoxy)ethanamine hydrochloride, 4-(2-Aminoethoxy)-1-nitrobenzene hydrochloride
AN-1140-5G
AN-1140-1G
COBBAN-1140-5GEA
375
EUR
COBBAN-1140-5G
COBBAN-1140-1G
4-Nitrophenethylamin Hydrochlorid
4-Nitrophenethylamin Hydrochlorid
Formel:
C₈H₁₁ClN₂O₃ Molecular Weight: 218.6400 g/mol |
MDL:
MFCD09475842 CAS-Nummer: 98395-62-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...