2-(Methylsulfonyl)anilin Hydrochlorid

Lieferant: CHEMPUR

JFD01521.250 JFD01521.1
CHMPJFD01521.250EA 0 EUR
CHMPJFD01521.250 MAYBJFD01521.1
2-(Methylsulfonyl)anilin Hydrochlorid
2-(Methylsulfonyl)anilin Hydrochlorid
Formel: C₇H₁₀ClNO₂S
MDL: MFCD00216484
CAS-Nummer: 205985-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR