4-(Phenylsulfonyl)anilin

Lieferant: FLUOROCHEM

019288-1G 019288-5G
FLUO019288-1GEA 116 EUR
FLUO019288-1G FLUO019288-5G
4-(Phenylsulfonyl)anilin
4-(Phenylsulfonyl)anilin
Formel: C₁₂H₁₁NO₂S
Molecular Weight: 233.2900 g/mol
MDL: MFCD00625703
CAS-Nummer: 7019-01-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR