(1H-Imidazol-4-yl)methanol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

219908-1G 219908-5G
SIAL219908-1GEA 30.2 EUR
SIAL219908-1G SIAL219908-5G
(1H-Imidazol-4-yl)methanol Hydrochlorid, Sigma-Aldrich®
(1H-Imidazol-4-yl)methanol Hydrochlorid
Formel: C₄H₇ClN₂O
Molecular Weight: 134,57 g/mol
MDL: MFCD00012697
CAS-Nummer: 32673-41-9
EINECS: 251-150-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR