5-Acetyluracil

Lieferant: BIONET RESEARCH

Synonyme: 5-Acetylpyrimidine-2,4(1H,3H)-dione

3E-902.500 3E-902.1 3E-902.5 3E-902.10
BIOT3E-902.500EA 0 EUR
BIOT3E-902.500 BIOT3E-902.1 BIOT3E-902.5 BIOT3E-902.10
5-Acetyluracil
5-Acetyluracil
Formel: C₆H₆N₂O₃
Molecular Weight: 154.11 g/mol
MDL: MFCD00151968
CAS-Nummer: 6214-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR