2-(Methoxycarbonylmethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-(2-Methoxy-2-oxoethyl)benzeneboronic acid, pinacol ester
PN-6025-1G
PN-6025-250M
COBBPN-6025-1GEA
0
EUR
COBBPN-6025-1G
COBBPN-6025-250M
2-(Methoxycarbonylmethyl)phenylboronsäurepinakolester
2-(Methoxycarbonylmethyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₄ Molecular Weight: 276.1358 g/mol |
MDL:
MFCD12913983 CAS-Nummer: 956229-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...