3-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate
PN-5817-5G
PN-5817-1G
PN-5817-250M
COBBPN-5817-5GEA
0
EUR
COBBPN-5817-5G
COBBPN-5817-1G
COBBPN-5817-250M
3-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
3-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
Formel:
C₁₆H₂₃BO₄ |
MDL:
MFCD08704682 CAS-Nummer: 1198615-70-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...