4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid ethyl ester, Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-[(4-Ethoxycarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Ethoxycarbonylmethyl)phenylboronic acid Pinacol ester
PN-8932-5G
PN-8932-1G
COBBPN-8932-5GEA
0
EUR
COBBPN-8932-5G
COBBPN-8932-1G
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
Formel:
C₁₆H₂₃BO₄ |
MDL:
MFCD08704684 CAS-Nummer: 859169-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...