4-(Methoxycarbonylmethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-[(4-MethoxyCarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid methyll ester, Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, Methyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate, 4-(MethoxyCarbonylmethyl)phenylboronic acid Pinacol ester

FM-2401-5G FM-2401-1G
COBBFM-2401-5GEA 235 EUR
COBBFM-2401-5G COBBFM-2401-1G
4-(Methoxycarbonylmethyl)phenylboronsäurepinakolester
4-(Methoxycarbonylmethyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,1358 g/mol
MDL: MFCD09997797
CAS-Nummer: 454185-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR