3-(Boc-amino)-4-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Boc-amino)-4-methylbenzeneboronic acid, 3-BOC-amino-4-methylphenylboronic acid, (3-((tert-Butoxycarbonyl)amino)-4-methylphenyl)boronic acid

BB-2483-5G BB-2483-1G
COBBBB-2483-5GEA 1310 EUR
COBBBB-2483-5G COBBBB-2483-1G
3-(Boc-amino)-4-methylphenylboronsäure
3-(Boc-amino)-4-methylphenylboronsäure
Formel: C₁₂H₁₈BNO₄
Molecular Weight: 251.09 g/mol
MDL: MFCD04115654
CAS-Nummer: 850568-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR