4-(Ethoxycarbonylmethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-[(Ethoxycarbonyl)methyl]phenylboronic acid, (4-(2-Ethoxy-2-oxoethyl)phenyl)boronic acid
BB-9006-1G
BB-9006-250M
COBBBB-9006-1GEA
0
EUR
COBBBB-9006-1G
COBBBB-9006-250M
4-(Ethoxycarbonylmethyl)phenylboronsäure
4-(Ethoxycarbonylmethyl)phenylboronsäure
Formel:
C₁₀H₁₃BO₄ Molecular Weight: 208.200 g/mol |
MDL:
MFCD08704683 CAS-Nummer: 92243-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...