2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Methyl-5-(piperidin-1-ylsulphonyl)phenyl)boronic acid
BB-2909-5G
BB-2909-1G
COBBBB-2909-5GEA
1690
EUR
COBBBB-2909-5G
COBBBB-2909-1G
2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronsäure
2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronsäure
Formel:
C₁₂H₁₈BNO₄S Molecular Weight: 283.1500 g/mol |
MDL:
MFCD08056364 CAS-Nummer: 871333-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...