2-Methyl-4-(piperidin-1-ylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Methyl-4-(piperidin-1-ylsulphonyl)phenyl)boronic acid

BB-6154-5G BB-6154-1G BB-6154-250M
COBBBB-6154-5GEA 0 EUR
COBBBB-6154-5G COBBBB-6154-1G COBBBB-6154-250M
2-Methyl-4-(piperidin-1-ylsulfonyl)phenylboronsäure
2-Methyl-4-(piperidin-1-ylsulfonyl)phenylboronsäure
Formel: C₁₂H₁₈BNO₄S
Molecular Weight: 283.1500 g/mol
MDL: MFCD13195672
CAS-Nummer: 1217501-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR