5-Brom-2-methyl-4-thiocyanatoanilin
Lieferant: COMBI-BLOCKS
AN-1400-1G
AN-1400-5G
COBBAN-1400-1GEA
98.3
EUR
COBBAN-1400-1G
COBBAN-1400-5G
5-Brom-2-methyl-4-thiocyanatoanilin
5-Brom-2-methyl-4-thiocyanatoanilin
Formel:
C₈H₇BrN₂S |
MDL:
MFCD11855817 CAS-Nummer: 1081803-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...