4-Brom-N,2-dimethylanilin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-N,2-dimethylaniline

HC-2987-5G HC-2987-1G
COBBHC-2987-5GEA 656 EUR
COBBHC-2987-5G COBBHC-2987-1G
4-Brom-N,2-dimethylanilin
4-Brom-N,2-dimethylanilin
Formel: C₈H₁₀BrN
Molecular Weight: 200,800 g/mol
MDL: MFCD11151443
CAS-Nummer: 59557-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR