3-Brom-N,4-dimethylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2873-5G HC-2873-1G
COBBHC-2873-5GEA 520 EUR
COBBHC-2873-5G COBBHC-2873-1G
3-Brom-N,4-dimethylanilin Hydrochlorid
3-Brom-N,4-dimethylanilin Hydrochlorid
Formel: C₈H₁₁BrClN
MDL: MFCD12756443
CAS-Nummer: 1187386-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR