5-Chlor-4-methoxy-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5415-1G BB-5415-250M
COBBBB-5415-1GEA 864 EUR
COBBBB-5415-1G COBBBB-5415-250M
5-Chlor-4-methoxy-2-methylphenylboronsäure
5-Chlor-4-methoxy-2-methylphenylboronsäure
Formel: C₈H₁₀BClO₃
MDL: MFCD09054938
CAS-Nummer: 511295-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR