2-((2-Chlor-5-(trifluormethyl)phenoxy)methyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-((2-Chloro-5-(trifluoromethyl)phenoxy)methyl)phenyl)boronic acid

BB-8067-1G BB-8067-250M
COBBBB-8067-1GEA 314 EUR
COBBBB-8067-1G COBBBB-8067-250M
2-((2-Chlor-5-(trifluormethyl)phenoxy)methyl)phenylboronsäure
2-((2-Chlor-5-(trifluormethyl)phenoxy)methyl)phenylboronsäure
Formel: C₁₄H₁₁BClF₃O₃
Molecular Weight: 330,5000 g/mol
MDL: MFCD07369758
CAS-Nummer: 849062-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR