4-((2-Chlor-5-(trifluormethyl)phenoxy)methyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-((2-Chloro-5-(trifluoromethyl)phenoxy)methyl)phenyl)boronic acid
BB-8082-1G
BB-8082-250M
COBBBB-8082-1GEA
330
EUR
COBBBB-8082-1G
COBBBB-8082-250M
4-((2-Chlor-5-(trifluormethyl)phenoxy)methyl)phenylboronsäure
4-((2-Chlor-5-(trifluormethyl)phenoxy)methyl)phenylboronsäure
Formel:
C₁₄H₁₁BClF₃O₃ Molecular Weight: 330.49 g/mol |
MDL:
MFCD09038412 CAS-Nummer: 849062-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...