3-Methyl-4-nitrobenzamid

Lieferant: CHEMPUR

TL00718.1 TL00718.10 TL00718.50
CHMPTL00718.1EA 0 EUR
CHMPTL00718.1 CHMPTL00718.10 CHMPTL00718.50
3-Methyl-4-nitrobenzamid
3-Methyl-4-nitrobenzamid
Formel: C₈H₈N₂O₃
Molecular Weight: 180,16 g/mol
MDL: MFCD00031468
CAS-Nummer: 99584-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR