Methyl-3-methylsalicylat

Lieferant: CHEMPUR

Synonyme: Methyl 2-hydroxy-3-methylbenzoate

SPB07565.250 SPB07565.1
CHMPSPB07565.250EA 0 EUR
CHMPSPB07565.250 MAYBSPB07565.1
Methyl-3-methylsalicylat
Methyl-3-methylsalicylat
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00020051
CAS-Nummer: 23287-26-5
EINECS: 245-557-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR